Assay Details
Target Gene Details
Entrez Gene ID: | 56095 |
Gene Name: | FtsJ homolog 3 (E. coli) |
Gene Aliases: |
AA537063, AU045295, C79843, D11Ertd400e, Epcs3 |
Location: |
Chr.11:106249144-106255802 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ftsj3 | NM_025310.2 | 14 | 1447 | NP_079586.1 |
| AK004996.2 | 14 | 1447 | BAB23730.1 | |
| AK031514.1 | 14 | 1434 | ||
| AK145490.1 | 14 | 1472 | BAE26467.1 | |
| BC012281.1 | 14 | 1444 | AAH12281.1 |

Genomic Map