Assay Details
Target Gene Details
Entrez Gene ID: | 56095 |
Gene Name: | FtsJ homolog 3 (E. coli) |
Gene Aliases: |
AA537063, AU045295, C79843, D11Ertd400e, Epcs3 |
Location: |
Chr.11:106249144-106255802 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ftsj3 | NM_025310.2 | 7 | 525 | NP_079586.1 |
AK004996.2 | 7 | 525 | BAB23730.1 | |
AK031514.1 | 7 | 512 | ||
AK145490.1 | 7 | 550 | BAE26467.1 | |
BC012281.1 | 7 | 522 | AAH12281.1 |
