Assay Details
Target Gene Details
Entrez Gene ID: | 56077 |
Gene Name: | diacylglycerol kinase, epsilon |
Gene Aliases: |
C87606, DAGK6, DGK |
Location: |
Chr.11:89037582-89060763 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgke | NM_019505.3 | NP_062378.1 | ||
| AF136744.1 | AAD45665.1 | |||
| AK049335.1 | ||||
| AK147841.1 |
More Information
Set Membership: |
|

Genomic Map