Assay Details
Target Gene Details
Entrez Gene ID: | 56077 |
Gene Name: | diacylglycerol kinase, epsilon |
Gene Aliases: |
C87606, DAGK6, DGK |
Location: |
Chr.11:89037582-89060763 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgke | NM_019505.3 | 4 | 1178 | NP_062378.1 |
| AF136744.1 | 4 | 815 | AAD45665.1 | |
| AK049335.1 | 4 | 978 | ||
| AK147841.1 | 4 | 1179 | ||
| BB635763.1 | 4 | 582 |
More Information
Set Membership: |
|

Genomic Map