Assay Details
Target Gene Details
Entrez Gene ID: | 52469 |
Gene Name: | cytochrome C oxidase assembly factor 3 |
Gene Aliases: |
1810033A19Rik, Ccdc56, D11Ertd99e, HSPC009 |
Location: |
Chr.11:101277970-101278948 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Coa3 | NM_026618.2 | 1 | 72 | NP_080894.1 |
| AK003955.1 | 1 | 70 | ||
| AK019025.1 | 1 | 74 | BAB31515.1 | |
| BC026206.1 | 1 | 66 | AAH26206.1 |
More Information
Set Membership: |
|

Genomic Map