Assay Details
Target Gene Details
Entrez Gene ID: | 69802 |
Gene Name: | cytochrome c oxidase assembly protein 11 |
Gene Aliases: |
2010004I09Rik |
Location: |
Chr.11:90638184-90645977 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox11 | NM_199008.2 | NP_950173.1 | ||
| AK008096.1 | ||||
| AK143937.1 | BAE25618.1 | |||
| AK172067.1 | ||||
| BC061233.1 | AAH61233.1 |
More Information
Set Membership: |
|

Genomic Map