Assay Details
Target Gene Details
Entrez Gene ID: | 14387 |
Gene Name: | glucosidase, alpha, acid |
Gene Aliases: |
E430018M07Rik |
Location: |
Chr.11:119267967-119285698 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gaa | NM_001159324.1 | 12 | 1860 | NP_001152796.1 |
| NM_008064.3 | 12 | 1950 | NP_032090.3 | |
| AK052211.1 | 12 | 1931 | BAC34888.1 | |
| AK088481.1 | 12 | 1926 | BAC40382.1 | |
| AK139333.1 | 12 | 1898 | BAE23960.1 | |
| AK140042.1 | 12 | 1918 | ||
| AK141546.1 | 12 | 1933 | ||
| AK145631.1 | 11 | 1366 | ||
| AK146538.1 | 12 | 1854 | BAE27243.1 | |
| AK150970.1 | 12 | 1908 | BAE30001.1 | |
| AK191262.1 | 1 | 44 | ||
| BC010210.1 | 12 | 1914 | AAH10210.1 | |
| U49351.1 | 12 | 1802 | AAB06943.1 |
More Information
Set Membership: |
|

Genomic Map