Assay Details
Target Gene Details
Entrez Gene ID: | 67181 |
Gene Name: | CTD nuclear envelope phosphatase 1 |
Gene Aliases: |
2610507E10Rik, Dullard |
Location: |
Chr.11:69981168-69990601 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctdnep1 | NM_026017.2 | NP_080293.1 | ||
| AK012063.1 | ||||
| AK076150.1 | ||||
| AK164602.1 | BAE37845.1 | |||
| BC018265.1 | AAH18265.1 |

Genomic Map