Assay Details
Target Gene Details
Entrez Gene ID: | 13681 |
Gene Name: | eukaryotic translation initiation factor 4A1 |
Gene Aliases: |
BM-010, Ddx2a, Eif4 |
Location: |
Chr.11:69666936-69672423 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |

Genomic Map