Assay Details
Target Gene Details
Entrez Gene ID: | 14387 |
Gene Name: | glucosidase, alpha, acid |
Gene Aliases: |
E430018M07Rik |
Location: |
Chr.11:119267967-119285698 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gaa | NM_001159324.1 | 17 | 2538 | NP_001152796.1 |
| NM_008064.3 | 17 | 2628 | NP_032090.3 | |
| AK052211.1 | 17 | 2609 | BAC34888.1 | |
| AK088481.1 | 17 | 2604 | BAC40382.1 | |
| AK139333.1 | 17 | 2576 | BAE23960.1 | |
| AK141546.1 | 17 | 2611 | ||
| AK145631.1 | 16 | 2044 | ||
| AK146538.1 | 17 | 2532 | BAE27243.1 | |
| AK150970.1 | 17 | 2586 | BAE30001.1 | |
| BC010210.1 | 17 | 2592 | AAH10210.1 | |
| U49351.1 | 17 | 2480 | AAB06943.1 |
More Information
Set Membership: |
|

Genomic Map