Assay Details
Target Gene Details
Entrez Gene ID: | 16191 |
Gene Name: | interleukin 5 |
Gene Aliases: |
Il-5 |
Location: |
Chr.11:53719364-53725623 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il5 | NM_010558.1 | NP_034688.1 | ||
BC125366.1 | ||||
BC132066.1 | ||||
X04601.1 | CAA28266.1 | |||
X06270.1 | CAA29606.1 |
