Assay Details
Target Gene Details
Entrez Gene ID: | 217038 |
Gene Name: | mitochondrial rRNA methyltransferase 1 homolog (S. cerevisiae) |
Gene Aliases: |
A530065E19Rik, ENSMUSG00000054212 |
Location: |
Chr.11:84813061-84819515 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrm1 | NM_145433.1 | NP_663408.2 | ||
| AK156416.1 | BAE33706.1 | |||
| AK163986.1 | BAE37571.1 | |||
| AK164059.1 | BAE37607.1 | |||
| BC005625.1 | AAH05625.2 |
More Information
Set Membership: |
|

Genomic Map