Assay Details
Target Gene Details
Entrez Gene ID: | 217214 |
Gene Name: | N-acetylglutamate synthase |
Gene Aliases: |
1700120E20Rik, argA |
Location: |
Chr.11:102145513-102149528 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nags | NM_145829.2 | NP_665828.1 | ||
| AF462069.1 | AAL86770.1 | |||
| AI415708.1 | ||||
| AJ489814.1 | CAD34015.1 | |||
| AK075765.1 | BAC35941.1 | |||
| BC057990.1 | AAH57990.1 |

Genomic Map