Assay Details
Target Gene Details
Entrez Gene ID: | 17168 |
Gene Name: | nitrogen permease regulator-like 3 |
Gene Aliases: |
Aag, CGTHBA, HS-26, HS-40, Mare, Phg, Prox1, m(alpha)RE |
Location: |
Chr.11:32231963-32267707 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nprl3 | NM_001284359.1 | NP_001271288.1 | ||
NM_001284360.1 | NP_001271289.1 | |||
NM_181569.3 | NP_853547.1 | |||
NR_104306.1 | ||||
AK036405.1 | BAC29415.1 | |||
AK036633.1 | BAC29512.1 | |||
AK039600.1 | ||||
AK047190.1 | BAC32985.1 | |||
AK081902.1 | BAC38366.1 | |||
AK154884.1 | ||||
AK184012.1 | ||||
BC064471.1 | AAH64471.1 |
