Assay Details
Target Gene Details
Entrez Gene ID: | 15254 |
Gene Name: | histidine triad nucleotide binding protein 1 |
Gene Aliases: |
AA673479, Hint, Ipk1, PKCI-1, PRKCNH1 |
Location: |
Chr.11:54866438-54870496 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hint1 | NM_008248.2 | 1 | 81 | NP_032274.1 |
| AK002965.2 | 1 | 81 | BAB22484.1 | |
| AK012433.1 | 1 | 87 | BAB28235.1 | |
| BC070415.1 | 1 | 126 | AAH70415.1 | |
| BC080296.1 | 1 | 65 | AAH80296.1 | |
| U60001.1 | 1 | 51 | AAC71076.1 |
More Information
Set Membership: |
|

Genomic Map