Assay Details
Target Gene Details
Entrez Gene ID: | 67036 |
Gene Name: | mitochondrial ribosomal protein L45 |
Gene Aliases: |
2600005P05Rik, AU043542, R75241 |
Location: |
Chr.11:97315716-97329920 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl45 | NM_025927.4 | NP_080203.1 | ||
| AK011163.1 | BAB27440.1 | |||
| AK011605.1 | BAB27727.1 | |||
| AK081922.1 | BAC38375.1 | |||
| AK157771.1 | BAE34190.1 | |||
| AK198473.1 | ||||
| BC020130.1 | AAH20130.1 | |||
| BY263659.1 |
More Information
Set Membership: |
|

Genomic Map