Assay Details
Target Gene Details
Entrez Gene ID: | 17428 |
Gene Name: | max binding protein |
Gene Aliases: |
Rox, bHLHd3 |
Location: |
Chr.11:74830796-74845725 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mnt | NM_010813.3 | 4 | 1163 | NP_034943.3 |
| AK139743.1 | 3 | 185 | ||
| AK147933.1 | 4 | 1147 | BAE28239.1 | |
| BC054534.1 | 4 | 1089 | AAH54534.1 | |
| U77356.1 | 4 | 1163 | AAB38687.1 | |
| Y07609.1 | 4 | 1109 | CAA68878.1 |
More Information
Set Membership: |
|

Genomic Map