Assay Details
Target Gene Details
Entrez Gene ID: | 64656 |
Gene Name: | mitochondrial ribosomal protein S23 |
Gene Aliases: |
D11Bwg1153e, MRP-S23, Rpms23, S23mt |
Location: |
Chr.11:88194106-88211507 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps23 | NM_001291270.1 | NP_001278199.1 | ||
| NM_001291271.1 | NP_001278200.1 | |||
| NM_001291272.1 | NP_001278201.1 | |||
| NM_001291273.1 | NP_001278202.1 | |||
| NM_024174.6 | NP_077136.2 | |||
| NR_111902.1 | ||||
| AK009880.1 | ||||
| AK014375.2 | BAB29307.2 | |||
| AK168051.1 | BAE40032.1 | |||
| AK168277.1 | BAE40223.1 | |||
| BC002155.1 | AAH02155.1 | |||
| BC019980.1 | AAH19980.1 | |||
| BY238199.1 |
More Information
Set Membership: |
|

Genomic Map