Assay Details
Target Gene Details
Entrez Gene ID: | 64660 |
Gene Name: | mitochondrial ribosomal protein S24 |
Gene Aliases: |
3110030K20Rik, AI414579, Rpms24 |
Location: |
Chr.11:5703983-5707717 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps24 | NM_026080.2 | NP_080356.1 | ||
| AK003117.1 | BAB22577.1 | |||
| AK007338.1 | BAE43203.1 | |||
| AK010590.1 | BAB27047.1 | |||
| AK012461.1 | ||||
| AK014099.1 | BAB29155.1 | |||
| BC029262.1 | AAH29262.1 | |||
| BY100364.1 |
More Information
Set Membership: |
|

Genomic Map