Assay Details
Target Gene Details
Entrez Gene ID: | 20185 |
Gene Name: | nuclear receptor co-repressor 1 |
Gene Aliases: |
5730405M06Rik, A230020K14Rik, N-CoR, RIP13, Rxrip13, mKIAA1047 |
Location: |
Chr.11:62315643-62457332 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncor1 | NM_001252313.1 | 21 | 3072 | NP_001239242.1 |
| NM_011308.3 | 21 | 2922 | NP_035438.3 | |
| AB093281.1 | 4 | 950 | BAC41465.1 | |
| AK137695.1 | 11 | 1774 | BAE23465.1 | |
| BC086657.1 | 21 | 2888 | AAH86657.1 | |
| U35312.1 | 20 | 3115 | AAB17125.1 |
More Information
Set Membership: |
|

Genomic Map