Assay Details
Target Gene Details
Entrez Gene ID: | 14783 |
Gene Name: | growth factor receptor bound protein 10 |
Gene Aliases: |
5730571D09Rik, AI325020, Meg1, mKIAA0207 |
Location: |
Chr.11:11930499-12037422 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grb10 | NM_010345.4 | 1 | 106 | NP_034475.2 |
| AF022072.1 | 1 | 80 | AAB72103.1 | |
| AK012646.1 | 1 | 86 | ||
| AK030727.1 | 1 | 41 | BAC27100.1 | |
| AK032927.1 | 1 | 87 | BAC28088.1 | |
| AK163841.1 | 1 | 87 | BAE37514.1 | |
| BC053842.1 | 1 | 73 | AAH53842.1 | |
| FJ425892.1 | 1 | 113 |
More Information
Set Membership: |
|

Genomic Map