Assay Details
Target Gene Details
Entrez Gene ID: | 76626 |
Gene Name: | musashi RNA-binding protein 2 |
Gene Aliases: |
1700105C15Rik, AI451722, AI563628, AW489193, Msi2h |
Location: |
Chr.11:88339382-88718267 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msi2 | NM_001201341.1 | NP_001188270.1 | ||
| NM_054043.3 | NP_473384.1 | |||
| AK031365.1 | BAC27370.1 | |||
| AK049637.1 | BAC33851.1 | |||
| AK049688.1 | BAC33873.1 | |||
| AK051269.1 | BAC34584.1 | |||
| AK153928.1 | BAE32260.1 | |||
| AK169846.1 | BAE41407.1 | |||
| BC109368.1 | AAI09369.1 | |||
| BC111809.1 | AAI11810.1 |
More Information
Set Membership: |
|

Genomic Map