Assay Details
Target Gene Details
Entrez Gene ID: | 20185 |
Gene Name: | nuclear receptor co-repressor 1 |
Gene Aliases: |
5730405M06Rik, A230020K14Rik, N-CoR, RIP13, Rxrip13, mKIAA1047 |
Location: |
Chr.11:62315643-62457332 on Build GRCm38 |
Assay Gene Location: | Within Exon 42 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncor1 | NM_001252313.1 | 40 | 6581 | NP_001239242.1 |
| NM_011308.3 | 40 | 6380 | NP_035438.3 | |
| AB093281.1 | 22 | 4321 | BAC41465.1 | |
| AK038502.1 | 1 | 1701 | ||
| AK137632.1 | 14 | 2699 | BAE23439.1 | |
| BC086657.1 | 40 | 6346 | AAH86657.1 | |
| L78294.1 | 19 | 3385 | AAL40135.1 | |
| U22016.1 | 6 | 1135 | AAC52168.1 | |
| U35312.1 | 39 | 6624 | AAB17125.1 |
More Information
Set Membership: |
|

Genomic Map