Assay Details
Target Gene Details
Entrez Gene ID: | 74302 |
Gene Name: | myotubularin related protein 3 |
Gene Aliases: |
1700092A20Rik, AI255150, AW557713, FYVE-DSP1, ZFYVE10, mKIAA0371 |
Location: |
Chr.11:4480868-4594815 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtmr3 | NM_028860.2 | 12 | 1511 | NP_083136.2 |
| AK141138.1 | 12 | 1491 | ||
| BC032166.1 | 12 | 1494 | AAH32166.1 | |
| BC081544.1 | 12 | 1495 | AAH81544.1 | |
| CB527330.1 | 3 | 275 | ||
| CV557784.1 |
More Information
Set Membership: |
|

Genomic Map