Assay Details
Target Gene Details
Entrez Gene ID: | 15184 |
Gene Name: | histone deacetylase 5 |
Gene Aliases: |
AI426555, Hdac4, mHDA1, mKIAA0600 |
Location: |
Chr.11:102195747-102230186 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hdac5 | NM_001077696.1 | 7 | 1016 | NP_001071164.1 |
| NM_001284248.1 | 9 | 1528 | NP_001271177.1 | |
| NM_001284249.1 | 8 | 1125 | NP_001271178.1 | |
| NM_001284250.1 | 7 | 1013 | NP_001271179.1 | |
| NM_010412.3 | 7 | 1013 | NP_034542.3 | |
| AF006602.2 | 6 | 729 | AAD09834.2 | |
| AF207748.1 | 6 | 648 | AAF31418.1 | |
| AK122325.2 | 2 | 52 | BAC65607.2 | |
| AK139202.1 | 8 | 1099 | ||
| AK145972.1 | 7 | 979 | ||
| AK146483.1 | 7 | 952 | BAE27204.1 | |
| AK155335.1 | 7 | 987 | BAE33201.1 | |
| AK156008.1 | 4 | 1264 | ||
| AK157159.1 | 4 | 1264 | ||
| AK191734.1 | ||||
| AK219061.1 | ||||
| BC060609.1 | 6 | 807 | AAH60609.1 |
More Information
Set Membership: |
|

Genomic Map