Assay Details
Target Gene Details
Entrez Gene ID: | 16399 |
Gene Name: | integrin alpha 2b |
Gene Aliases: |
AI172977, CD41, CD41B, GpIIb, alphaIIb |
Location: |
Chr.11:102453297-102469883 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itga2b | NM_010575.2 | 7 | 902 | NP_034705.2 |
| AF166384.1 | 7 | 780 | AAF43997.1 | |
| AF170316.1 | 7 | 763 | AAD56216.1 | |
| AK048700.1 | 3 | 257 | BAC33425.1 | |
| AK140749.1 | 3 | 235 | ||
| AK142289.1 | 7 | 764 | BAE25013.1 | |
| AK154619.1 | 7 | 903 | BAE32718.1 | |
| AK156872.1 | 7 | 765 | ||
| AK157012.1 | 7 | 764 | ||
| AK158443.1 | 7 | 749 | ||
| BC120493.1 | 7 | 752 | ||
| BC137570.1 | 7 | 752 |
More Information
Set Membership: |
|

Genomic Map