Assay Details
Target Gene Details
Entrez Gene ID: | 16878 |
Gene Name: | leukemia inhibitory factor |
Gene Aliases: |
- |
Location: |
Chr.11:4257568-4272514 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lif | NM_008501.2 | NP_032527.1 | ||
| BY207460.1 |
More Information
Set Membership: |
|

Genomic Map