Assay Details
Target Gene Details
Entrez Gene ID: | 98238 |
Gene Name: | leucine rich repeat containing 59 |
Gene Aliases: |
AA959742, C78668 |
Location: |
Chr.11:94629824-94645216 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrc59 | NM_133807.1 | NP_598568.1 | ||
| AK053450.1 | BAC35390.1 | |||
| AK159374.1 | BAE35030.1 | |||
| AK159552.1 | BAE35177.1 | |||
| AK166576.1 | BAE38865.1 | |||
| AK167604.1 | BAE39660.1 | |||
| AK169250.1 | BAE41014.1 | |||
| AK169390.1 | BAE41137.1 | |||
| AK183165.1 | ||||
| BC006877.1 | AAH06877.1 | |||
| BC066172.1 | AAH66172.1 |
More Information
Set Membership: |
|

Genomic Map