Assay Details
Target Gene Details
Entrez Gene ID: | 50997 |
Gene Name: | membrane protein, palmitoylated 2 (MAGUK p55 subfamily member 2) |
Gene Aliases: |
D11Bwg0652e, Dlg2, Dlgh2, Pals4 |
Location: |
Chr.11:102057016-102088636 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpp2 | NM_016695.3 | NP_057904.1 | ||
| AF162685.1 | AAD44342.1 | |||
| AK161577.1 | BAE36473.1 | |||
| AK172253.1 | BAE42909.1 | |||
| BC053026.1 | AAH53026.1 |
More Information
Set Membership: |
|

Genomic Map