Assay Details
Target Gene Details
Entrez Gene ID: | 17913 |
Gene Name: | myosin IC |
Gene Aliases: |
C80397, MMIb, MYO1E, NMI, mm1beta, myr2 |
Location: |
Chr.11:75650493-75674643 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myo1c | NM_008659.3 | 1 | 115 | NP_032685.1 |
| AK004743.1 | BAB23524.1 | |||
| AK155530.1 | 1 | 91 | BAE33311.1 | |
| BC021481.1 | AAH21481.1 | |||
| BY310607.1 | 1 | 115 | ||
| X99638.1 | 1 | 74 | CAA67956.1 |
More Information
Set Membership: |
|

Genomic Map