Assay Details
Target Gene Details
Entrez Gene ID: | 78304 |
Gene Name: | N(alpha)-acetyltransferase 38, NatC auxiliary subunit |
Gene Aliases: |
1500034E06Rik, Lsmd1 |
Location: |
Chr.11:69395791-69396671 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Naa38 | NM_030083.2 | NP_084359.1 | ||
| AK018782.1 | BAB31406.1 | |||
| BC048499.1 | ||||
| BC062267.1 | AAH62267.1 |
Target Gene Details
Entrez Gene ID: | 67020 |
Gene Name: | transmembrane protein 88 |
Gene Aliases: |
2600017H02Rik |
Location: |
Chr.11:69396516-69398234 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem88 | NM_025915.4 | 2 | 1461 | NP_080191.3 |
| AK045149.1 | 2 | 1462 | BAC32239.1 | |
| AK161789.1 | 2 | 1462 | BAE36575.1 |
More Information
Set Membership: |
|

Genomic Map