Assay Details
Target Gene Details
Entrez Gene ID: | 13384 |
Gene Name: | membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3) |
Gene Aliases: |
6430514B01, Dlgh3 |
Location: |
Chr.11:101999652-102027048 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpp3 | NM_007863.2 | 10 | 1022 | NP_031889.2 |
| AF079366.1 | 10 | 973 | AAD12762.1 | |
| AK032270.1 | 9 | 2296 | ||
| AY236514.1 | 9 | 856 | AAO65586.1 | |
| BC062642.1 | 10 | 990 | AAH62642.1 |
More Information
Set Membership: |
|

Genomic Map