Assay Details
Target Gene Details
Entrez Gene ID: | 17913 |
Gene Name: | myosin IC |
Gene Aliases: |
C80397, MMIb, MYO1E, NMI, mm1beta, myr2 |
Location: |
Chr.11:75650493-75674643 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myo1c | NM_001080774.1 | 25 | 2685 | NP_001074243.1 |
| NM_001080775.1 | 25 | 2678 | NP_001074244.1 | |
| NM_008659.3 | 25 | 2648 | NP_032685.1 | |
| AK004743.1 | 25 | 2577 | BAB23524.1 | |
| AK154294.1 | 25 | 2544 | BAE32495.1 | |
| AK155530.1 | 25 | 2624 | BAE33311.1 | |
| AK166638.1 | 25 | 2684 | ||
| AK171107.1 | 25 | 2679 | BAE42253.1 | |
| AY007255.1 | 25 | 2534 | AAG02570.1 | |
| BC021481.1 | 25 | 2589 | AAH21481.1 | |
| U96723.1 | 24 | 2472 | AAC53264.1 |
More Information
Set Membership: |
|

Genomic Map