Assay Details
Target Gene Details
Entrez Gene ID: | 66809 |
Gene Name: | keratin 20 |
Gene Aliases: |
9030623C06Rik, CK20, K20, KRT21 |
Location: |
Chr.11:99428403-99438153 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt20 | NM_023256.1 | 4 | 790 | NP_075745.1 |
| AF473907.1 | 4 | 767 | AAL82480.1 | |
| AK018567.1 | 4 | 790 | BAB31280.1 | |
| BC057172.1 | 4 | 773 | AAH57172.1 |

Genomic Map