Assay Details
Target Gene Details
Entrez Gene ID: | 13384 |
Gene Name: | membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3) |
Gene Aliases: |
6430514B01, Dlgh3 |
Location: |
Chr.11:101999652-102027048 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpp3 | NM_007863.2 | NP_031889.2 | ||
| AF079366.1 | AAD12762.1 | |||
| AK032270.1 | ||||
| AY236514.1 | AAO65586.1 | |||
| BC062642.1 | AAH62642.1 |
More Information
Set Membership: |
|

Genomic Map