Assay Details
Target Gene Details
Entrez Gene ID: | 78455 |
Gene Name: | helicase with zinc finger domain |
Gene Aliases: |
3110078M01Rik, 9430093I07Rik, 9630002H22Rik, AI851979, DRHC |
Location: |
Chr.11:107546958-107693826 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 20 - Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Helz | NM_198298.1 | NP_938040.1 | ||
| AB093209.1 | BAC41393.1 | |||
| AK033094.1 | BAC28150.1 | |||
| AK044226.1 | ||||
| BC076626.1 | AAH76626.1 |

Genomic Map