Assay Details
Target Gene Details
Entrez Gene ID: | 192775 |
Gene Name: | potassium voltage-gated channel, subfamily H (eag-related), member 6 |
Gene Aliases: |
m-erg2 |
Location: |
Chr.11:106007994-106035595 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnh6 | NM_001037712.1 | 7 | 1564 | NP_001032801.1 |
| AK038513.1 | 1 | 63 | BAC30022.1 | |
| BC109174.1 | 7 | 1564 | AAI09175.1 |
More Information
Set Membership: |
|

Genomic Map