Assay Details
Target Gene Details
Entrez Gene ID: | 70382 |
Gene Name: | potassium channel tetramerisation domain containing 2 |
Gene Aliases: |
2310012I15Rik |
Location: |
Chr.11:115420126-115431274 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kctd2 | NM_183285.3 | NP_899108.3 | ||
AA537633.1 | ||||
AK009318.1 | BAC25251.1 | |||
BC095988.1 | AAH95988.1 |
