Assay Details
Target Gene Details
Entrez Gene ID: | 192159 |
Gene Name: | pre-mRNA processing factor 8 |
Gene Aliases: |
AU019467, D11Bwg0410e, DBF3/PRP8, Prp8, Sfprp8l |
Location: |
Chr.11:75486777-75509447 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prpf8 | NM_138659.2 | 2 | 491 | NP_619600.2 |
| AB047391.1 | 2 | 217 | BAB32671.1 | |
| AK041017.1 | 3 | 258 | BAC30783.1 | |
| AK144232.1 | 2 | 416 | BAE25786.1 | |
| BC093481.1 | 2 | 421 | ||
| CJ120731.1 |
More Information
Set Membership: |
|

Genomic Map