Assay Details
Target Gene Details
Entrez Gene ID: | 20425 |
Gene Name: | serine hydroxymethyltransferase 1 (soluble) |
Gene Aliases: |
AI324848, AI385541, C81125, Shmt, mshmt, mshmt1, mshmt2 |
Location: |
Chr.11:60788892-60811265 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Shmt1 | NM_009171.2 | NP_033197.2 | ||
| AF237702.1 | AAK15040.1 | |||
| AK010439.1 | BAB26940.1 | |||
| BQ942592.1 | ||||
| CF159149.1 | ||||
| X94478.1 | CAA64225.1 | |||
| X94479.1 | CAA64226.1 |
More Information
Set Membership: |
|

Genomic Map