Assay Details
Target Gene Details
Entrez Gene ID: | 74467 |
Gene Name: | pseudouridylate synthase 10 |
Gene Aliases: |
2810013G11Rik, 4933435A13Rik, AU014648, C77560, Ccdc139 |
Location: |
Chr.11:23665354-23732876 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pus10 | NM_001033654.2 | NP_001028826.1 | ||
| NM_028304.2 | NP_082580.1 | |||
| NM_028956.4 | NP_083232.1 | |||
| AK012737.1 | BAB28439.1 | |||
| AK017063.1 | BAB30576.1 | |||
| AK028368.1 | BAC25908.1 | |||
| AK029866.1 | BAC26650.1 | |||
| AK088562.1 | BAC40426.1 | |||
| AK145055.1 | BAE26208.1 | |||
| BC025555.1 | AAH25555.1 |
More Information
Set Membership: |
|

Genomic Map