Assay Details
Target Gene Details
Entrez Gene ID: | 110880 |
Gene Name: | sodium channel, voltage-gated, type IV, alpha |
Gene Aliases: |
Nav1.4, SkM1, mH2 |
Location: |
Chr.11:106317545-106365552 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scn4a | NM_133199.2 | 11 | 1627 | NP_573462.2 |
| AJ278787.1 | 11 | 1627 | CAC17146.1 | |
| BC129805.1 | 11 | 1625 |
More Information
Set Membership: |
|

Genomic Map