Assay Details
Target Gene Details
Entrez Gene ID: | 18618 |
Gene Name: | phosphatidylethanolamine N-methyltransferase |
Gene Aliases: |
PEAMT, PEMT2, Pempt, Pempt2 |
Location: |
Chr.11:59970614-60046512 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pemt | NM_001290011.1 | 4 | 443 | NP_001276940.1 |
| NM_001290012.1 | 4 | 439 | NP_001276941.1 | |
| NM_001290013.1 | 4 | 377 | NP_001276942.1 | |
| NM_001290014.1 | NP_001276943.1 | |||
| NM_008819.3 | 4 | 477 | NP_032845.2 | |
| AK133744.1 | 4 | 439 | BAE21818.1 | |
| AY334571.1 | 4 | 336 | AAQ01190.1 | |
| BC026796.1 | 4 | 311 | AAH26796.1 | |
| BG084478.2 | 4 | 341 | ||
| BY251884.1 | 3 | 335 | ||
| CJ048474.1 | 4 | 439 |
More Information
Set Membership: |
|

Genomic Map