Assay Details
Target Gene Details
Entrez Gene ID: | 66249 |
Gene Name: | partner of NOB1 homolog (S. cerevisiae) |
Gene Aliases: |
1810003N24Rik, AA407421, AU015468, AU019495, AU042323, Emi3, Imi3 |
Location: |
Chr.11:17203198-17211589 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pno1 | NM_025443.2 | NP_079719.1 | ||
| AF349950.1 | AAK29665.1 | |||
| AK007324.1 | BAB24959.1 | |||
| AK009146.1 | BAB26104.1 | |||
| AK009896.1 | ||||
| AK017153.1 | ||||
| AK146152.1 | BAE26936.1 | |||
| AK165942.1 | BAE38474.1 | |||
| AK168177.1 | BAE40137.1 | |||
| AK205314.1 | ||||
| BC020037.1 | AAH20037.1 | |||
| BY034432.1 |

Genomic Map