Assay Details
Target Gene Details
Entrez Gene ID: | 215113 |
Gene Name: | solute carrier family 43, member 2 |
Gene Aliases: |
7630402D21Rik, BC042513, Lat4 |
Location: |
Chr.11:75530282-75577575 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc43a2 | NM_001199283.1 | 13 | 1820 | NP_001186212.1 |
NM_001199284.1 | 13 | 1602 | NP_001186213.1 | |
NM_173388.2 | 13 | 1565 | NP_775564.1 | |
AB120363.1 | 13 | 1598 | BAD91090.1 | |
AK143773.1 | 13 | 1663 | BAE25533.1 | |
AK158440.1 | 4 | 541 | BAE34511.1 | |
AK159144.1 | 13 | 1788 | BAE34852.1 | |
AK171104.1 | 13 | 1822 | ||
BC042513.1 | 13 | 1561 | AAH42513.1 |
