Assay Details
Target Gene Details
Entrez Gene ID: | 215113 |
Gene Name: | solute carrier family 43, member 2 |
Gene Aliases: |
7630402D21Rik, BC042513, Lat4 |
Location: |
Chr.11:75530282-75577575 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc43a2 | NM_001199283.1 | 2 | 445 | NP_001186212.1 |
NM_001199284.1 | 2 | 227 | NP_001186213.1 | |
NM_173388.2 | 2 | 190 | NP_775564.1 | |
AB120363.1 | 2 | 223 | BAD91090.1 | |
AK090207.1 | 1 | 524 | ||
AK143773.1 | 2 | 288 | BAE25533.1 | |
AK159144.1 | 2 | 413 | BAE34852.1 | |
AK165721.1 | 1 | 560 | ||
AK171104.1 | 2 | 445 | ||
AK172685.1 | 2 | 215 | ||
BC042513.1 | 2 | 186 | AAH42513.1 | |
BY202700.1 | 2 | 271 | ||
CD550550.1 | 2 | 186 |
