Assay Details
Target Gene Details
Entrez Gene ID: | 18436 |
Gene Name: | purinergic receptor P2X, ligand-gated ion channel, 1 |
Gene Aliases: |
AI323649, BB122383, P2x, Pdcd3, RP-2 |
Location: |
Chr.11:72999056-73015200 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2rx1 | NM_008771.3 | NP_032797.3 | ||
| AK035304.1 | BAC29024.1 | |||
| BC015084.1 | AAH15084.1 | |||
| X84896.1 | CAA59322.1 |
More Information
Set Membership: |
|

Genomic Map