Assay Details
Target Gene Details
Entrez Gene ID: | 103711 |
Gene Name: | pyridoxine 5'-phosphate oxidase |
Gene Aliases: |
AI415282 |
Location: |
Chr.11:96937816-96944019 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnpo | NM_134021.2 | NP_598782.1 | ||
| AK154004.1 | BAE32309.1 | |||
| AK154443.1 | BAE32590.1 | |||
| AK164667.1 | BAE37867.1 | |||
| BC010785.1 | AAH10785.1 | |||
| BC026564.1 | AAH26564.1 | |||
| CK625276.1 |
More Information
Set Membership: |
|

Genomic Map