Assay Details
Target Gene Details
Entrez Gene ID: | 52466 |
Gene Name: | solute carrier family 46, member 1 |
Gene Aliases: |
1110002C08Rik, D11Ertd18e, HCP1, Pcft |
Location: |
Chr.11:78465701-78471945 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc46a1 | NM_026740.2 | 4 | 1357 | NP_081016.2 |
| AK003278.1 | 4 | 1340 | BAB22685.1 | |
| AK170505.1 | 4 | 1350 | BAE41843.1 | |
| AK220201.1 | 4 | 1317 | BAD90126.1 | |
| BC024522.1 | 3 | 493 | AAH24522.1 | |
| BC057976.1 | 4 | 1342 | AAH57976.1 |

Genomic Map