Assay Details
Target Gene Details
Entrez Gene ID: | 103677 |
Gene Name: | Smg-6 homolog, nonsense mediated mRNA decay factor (C. elegans) |
Gene Aliases: |
AI317223, AU041178 |
Location: |
Chr.11:74925872-75164448 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smg6 | NM_001002764.1 | 14 | 3519 | NP_001002764.1 |
| AK129203.1 | 13 | 2138 | BAC98013.1 | |
| BC066040.1 | 14 | 3519 | AAH66040.1 |
More Information
Set Membership: |
|

Genomic Map